ADC® Replacement Bulbs And Valves
Supplier: AMERICAN DIAGNOSTIC CORP CP
Certificates
About this item
Fits most pocket, clock and mercury systems.
- Available in three sizes
- Fit many other brands
- Not manufactured using natural rubber latex
With three options to choose from its easy to replace a worn out bulb and valve on your sphygmomanometer. While any of these bulbs would make a great replacement, the adflow bulbs incorporate a screened end valve allowing for better airflow than a basic valve. The large adflow bulb allows for faster inflation, especially with larger bladders.
Frequently Bought Together