ContactUsLinkComponent
ADC® Diagnostix 5410 Diagnostic Set
Catalog # 470315-668
undefined
ADC® Diagnostix 5410 Diagnostic Set
Catalog # 470315-668
Supplier Number:  5410X
Specifications
  • Description:
    Diagnostix 3.5 V Portable Set
  • Cat. no.:
    470315-668
  • Voltage:
    3.5 V
Specifications
About this item
A great starter st with a rechargeable power supply.
  • Includes power handle, otoscope and ophthalmoscope
  • Storage case with formed liner for storage
Diagnostic EENT, 3.5 V instruments are among the most critical toolsets in a clinician's repertoire when performing a comprehensive physical exam. ADC Diagnostix-brand EENT instrument sets help practitioners perform fast, accurate, and efficient exams thanks to innovative optics, advanced lighting technologies, and long-lasting power sources. This combination of tools connects to a Li-ion battery handle. The handle plugs directly into an outlet for easy recharging and you can expect approximately 30 hours of use between charges. The included otoscope and ophthalmoscope heads attach securely to the handle and use halogen bulbs. Both the ophthalmoscope and the otoscope utilize advanced optical technology which creates a panoramic, magnified view that features twice the field of vision and 66% greater magnification (11 mm field of view, 5X magnification) compared to the standard ADC otoscope. Significantly improved view of the auditory canal and tympanic membrane.
Fiber-optic light transmission for an unobstructed view. Reflection-optimized, high-performance glass optics. Adjustable focal depth. Suitable for throat illumination and pneumatic otoscopy (insufflator sold separately). Compatible with Welch Allyn specula. The ophthalmoscope-theft locking feature is standard.