ADC® Proscope 5211 Otoscopes
Supplier: AMERICAN DIAGNOSTIC CORP CP
Certificates
About this item
A great place to start.
- C cell sized handle
- Includes 2.5 V Xenon bulb
- Includes zippered storage case
Want to start with an instrument you can easily expand as necessary? This proscope otoscope is ready for the challenge. The otoscope includes a C-cell base for power, with a strong 2.5 V Xenon bulb and a knurled finish for a secure grip. The otoscope head has a 3X magnifier, an insufflation port (bulb and connection not included) and is easily removed using a secure bayonet mount. The set includes three autoclavable specula, and you can additionally purchase
Accessories information: Does not include batteries.
Frequently Bought Together