ContactUsLinkComponent
VWR® Upright Freezer Drawer Rack for 96-Well Microtube Boxes
Catalog # 76051-838
Supplier:  VWR International
undefined
VWR® Upright Freezer Drawer Rack for 96-Well Microtube Boxes
Catalog # 76051-838
Supplier:  VWR International
Specifications
  • Width:
    5⁷⁄₁₆"
  • Capacity:
    24
  • Height:
    9¹¹⁄₁₆"
  • Length:
    21½"
  • Cat. no.:
    76051-838
Specifications
About this item
Designed for use with 96-well microtube boxes.
  • Made of corrosion resistant stainless steel
  • Security lock devices are available for each rack
Drawer racks allow easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.