VWR® Upright Freezer Drawer Rack for 96-Well Microtube Boxes
Catalog # 76051-748
Supplier: VWR International
Specifications
- Width:5⁷⁄₁₆"
- Capacity:9
- Height:7⁵⁄₁₆"
- Length:10¹⁵⁄₁₆"
- Cat. no.:76051-748
Specifications
About this item
Designed for use with 96-well microtube boxes.
- Made of corrosion resistant stainless steel
- Security lock devices are available for each rack
Drawer racks allow easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.