Axygen® MultiRack Maximum Recovery® Tips, Corning | Avantor
Axygen® MultiRack Maximum Recovery® Tips, Corning
Supplier:   Corning
Axygen® MultiRack Maximum Recovery® Tips, Corning
Supplier:  Corning
Certificates
Axygen® MultiRack Maximum Recovery® Tips, Corning
Axygen® MultiRack Maximum Recovery® Tips, Corning
4 Options Available Below
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack Maximum Recovery® tips are available with or without aerosol filters. Axygen® Maximum Recovery® technology creates a hydrophobic surface to improve accuracy and precision while reducing surface tension and sample wastage. They are specially designed for lab applications that demand complete release of a liquid solution, such as low concentration DNA, selected protein, and viscous liquids (glycerin, Triton-X and more). Axygen® MultiRack Maximum Recovery® tips are available in a wide range of tip sizes with filters and sterilization.
Ordering information: Axygen® MultiRack pipette tips are also available in bulk, hinged rack, filter tips, reload, and tipstation configurations.
Product Family Options
Informations sur le produit
  • Tip Style
    Volume
    Sterile
    Packed
    Availability
    Price
  • Standard Length
    1 - 200 µl
    +
    960 Tips/Pack, 5 Packs/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4,800
    Case of 4,800 - $477.75
    $477.75
    Prix promotionnel pour les nouveaux clients
    Case of 4,800
       $477.75
    Supplier Number: MR-200-L-R-S
  • Standard Length
    1 - 300 µl
    +
    960 Tips/Pack, 5 Packs/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4,800
    Case of 4,800 - $476.25
    $476.25
    Prix promotionnel pour les nouveaux clients
    Case of 4,800
       $476.25
    Supplier Number: MR-300-L-R-S
  • Extended Length
    0.1 - 10 µl
    +
    960 Tips/Pack, 5 Packs/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4,800
    Case of 4,800 - $477.75
    $477.75
    Prix promotionnel pour les nouveaux clients
    Case of 4,800
       $477.75
    Supplier Number: MR-10XT-L-R-S
  • Extended Length
    1 - 300 µl
    +
    768 Tips/Pack, 5 Packs/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 3,840
    Case of 3,840 - $635.00
    $635.00
    Case of 3,840
       $635.00
    Supplier Number: MR-300XT-L-R-S
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-744
    10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
    Log in pour la disponibilité
    Case of 5 - $133.50
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-746
    200 µl extended length, 300 µl extended length, and 1000 µl extended length tips
    Log in pour la disponibilité
    Case of 5 - $180.00