Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Description:Acrylamide bis-acrylamide
- Size:1 L
- Cat. no.:CA1710
- Supplier No.:1710
- Storage conditions:2...8 °C
- Form:Clear, colorless liquid
OmniPur® acrylamide bis-acrylamide, 29:1, 40% solution, is suitable for nucleic acid and protein separation sequencing. Premixed solutions offer standard ratios of acrylamide to bis-acrylamide for convenience and safety. The hazard of handling powdered acrylamide is eliminated with the 40% solution in water. Reconstitute with deionized H₂O to desired final percentage.
- Identity (IR-spectrum): conforms
- Water (H₂O): 1% max
Each pouch makes 1 L of 10X TBE buffer (1X = 0.089 M TRIS, 0.089 M borate, 0.002 M EDTA).