Axygen® Linear Stands for Four or Six Pipettors, Corning
: Corning
Universal linear stands for up to four or six pipettors.
Red model: For up to four single-channel pipettors or maximum two 8-channel or 12-channel pipettors.
Transparent model: For up to six single-channel pipettors or maximum three 8-channel or 12-channel pipettors.
- Catalog No:
- 76324-852
- 76324-848
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Warsaw
- I-RECs, ISO 14001 - Warsaw
Frequently Bought Together