Axygen® Multirack Bulk Tips, Corning
Supplier:   Corning
Axygen® Multirack Bulk Tips, Corning
Supplier:  Corning
Certificates
Axygen® Multirack Bulk Tips, Corning
Axygen® Multirack Bulk Tips, Corning
3 Options Available Below
Product Details & Documents
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an inter-changeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
Axygen® MultiRack bulk pipette tips are compatible with MultiRack racks and packaged in resealable polyethylene bags, non sterile.
 : Axygen® MultiRack pipette tips are also available in hinged rack, filter tips, reload, and tipstation configurations.
Product Family Options
Informations sur le produit
  • Tip Style
    Volume
    Packed
    Availability
    Price
  • Standard Length
    1 - 200 µl
    Bulk: 1000 Tips/Bag, 10 Bags/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10,000
    Case of 10,000 - $411.00
    $411.00
    Prix promotionnel pour les nouveaux clients
    Case of 10,000
       $411.00
    Supplier Number: MR-200
  • Extended Length
    0.1 - 10 µl
    Bulk: 1000 Tips/Bag, 10 Bags/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 10,000
    Case of 10,000 - $429.00
    $429.00
    Prix promotionnel pour les nouveaux clients
    Case of 10,000
       $429.00
    Supplier Number: MR-10XT
  • Extended Length
    50 - 1250 µl
    Bulk: 768 Tips/Bag, 10 Bags/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 7,680
    Case of 7,680 - $432.00
    $432.00
    Prix promotionnel pour les nouveaux clients
    Case of 7,680
       $432.00
    Supplier Number: MR-1000XT
Accessories
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-744
    10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
    Log in pour la disponibilité
    Case of 5 - $133.50
Informations sur le produit
  • Catalog #
    For Use With
    Availability
    Price
  • MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    76320-746
    200 µl extended length, 300 µl extended length, and 1000 µl extended length tips
    Log in pour la disponibilité
    Case of 5 - $180.00