ContactUsLinkComponent
SP Bel-Art Poxygrid® Poxygrid® Hybridization Bottle Rack, SP Scienceware, Bel-Art Products, a part of SP
Catalog # 76326-304
undefined
SP Bel-Art Poxygrid® Poxygrid® Hybridization Bottle Rack, SP Scienceware, Bel-Art Products, a part of SP
Catalog # 76326-304
Supplier Number:  F16964-0006
Specifications
  • Dimensions:
    13.7L×19.8W×13H cm (53/8×713/16×51/8")
  • Cat. no.:
    76326-304
  • No. of Places:
    6
Specifications
About this item
Heavy-duty rack protects your expensive bottles keeping them from rolling, tipping and striking against one another.
  • Epoxy-coated steel rods provide durability and easy cleaning
  • Steam autoclavable at 121 °C (250 °F)
  • Holes are 42 mm square shaped for holding 40 mm diameter bottles
Holds 150 and 300 mm long bottles with ample space in between to allow for the screw cap diameter and provide convenient handling.