Mounting Medium, Azer Scientific
Mounting Medium, Azer Scientific
Supplier:   Azer Scientific
Mounting Medium, Azer Scientific
Supplier:  Azer Scientific
Certificates
Mounting Medium, Azer Scientific
Mounting Medium, Azer Scientific
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Adhesive mounting medium with permanent clarity for long term slide storage.
  • Traditional formula or quick drying acrylic
  • Low viscosity for easy cover glass mounting
Non-aqueous S-mounting medium formula is compatible with xylene and most xylene substitutes, dries quickly and and will not crack or yellow over time.
Traditional mounting medium is soluble in xylene and toluene, dries optically clear and will resist photobleaching and stain fading.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • S-Mounting Medium
    4 oz. Dropper Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12 (1/Itm/Bx)
    Case of 12 (1/Itm/Bx) - $2,616.27
    $2,616.27
    Case of 12 (1/Itm/Bx)
       $2,616.27
    Supplier Number: ES723
  • S-Mounting Medium
    16 oz.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12 (1/Itm/Bx)
    Case of 12 (1/Itm/Bx) - $2,961.96
    $2,961.96
    Case of 12 (1/Itm/Bx)
       $2,961.96
    Supplier Number: ES724
  • Mounting Medium
    4 oz. Dropper Bottle
    Discontinued
    Supplier Number: -
  • Mounting Medium
    16 oz.
    Discontinued
    Supplier Number: -