FormaPad™ Neutralizing and Grossing Pads, Azer Scientific
Fournisseur: Azer Scientific
Certificats
À propos de cet article
Disposable grossing pads absorb and neutralize weak formaldehyde solutions.
- Pad surface will not shred or dull scalpel blades
- Quickly absorbs and neutralizes formalin
FormaPad™ pads are thick and strong enough to use as a grossing surface but can easily be cut down to size for safe transport of formalin filled specimen containers.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together