BioFit® Footrest with Chrome Metal Finish, BioFit Engineered Products
Catalog # 75821-288
Supplier: BioFit
Product available on GSA Advantage®
Specifications
- Description:BioFit Footrest
- Material:Chrome Metal
- Size:20W×11"D (Platform)
- Cat. no.:75821-288
Specifications
About this item
BioFit adjustable footrests can help enhance user comfort and performance. This model is ideal for use in office, laboratory, healthcare, technical, education, and industrial environments.
- Constructed of rugged 16-gauge steel tubing
- Fast, easy, and secure rack-and-pin height adjustment from 3 to 11"
- Non-skid platform angled at 10° slope
BioFit workplace furniture solutions are crafted with user comfort, health, and wellbeing in mind. Products meet or exceed rigorous industry standards and nearly all carry a 13-year warranty.
All BioFit products deliver proven performance and reliability. BIoFit company roots include over 70 year's background improving the workplace experience by providing ergonomic, LEED® compliant seating and more for healthcare, technology, education, lab, industrial, office and custom applications.
Caution: This product can expose users to chemicals including wood dust, which is known to the State of California to cause cancer. For more information go to www.p65warnings.ca.gov.