Tweezers, Peek Soft Replaceable Tip, Excelta Corp®
Supplier:   Excelta
Tweezers, Peek Soft Replaceable Tip, Excelta Corp®
Supplier:  Excelta
Certificates
Tweezers, Peek Soft Replaceable Tip, Excelta Corp®
Tweezers, Peek Soft Replaceable Tip, Excelta Corp®
3 Options Available Below
Product Details & Documents
Peek tweezers are designed for high temperature, are chemical resistant and will not scratch or marr delicate items.
  • High temperature resistant
  • Excellent chemical resistance
  • Soft tips won't scratch or marr
The Peek replaceable tip tweezers are made of a high temperature thermoplastic with excellent chemical resistance. The surface resistivity is 10⁴ to 10⁵ ohms/sq. They feature finger serrations for added control. The bodies are made of anti-magnetic stainless steel (316L). Replacement tips included 2 screws and 2 tips. They are excellent for applications requiring no metal-to-metal contact, avoiding scratching and marring.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Overall Length
    Tip Length
    Tip Style
    Tip Width
    Curvature
    Availability
    Price
  • Tweezers, replaceable tip, 5 star quality
    Peek tips
    125 mm (5")
    0.5 mm (0.02")
    Straight Very Fine Replaceable Points
    0.02" (0.5 mm)
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $230.49
    $230.49
    Each
       $230.49
    Supplier Number: 162A-RT
  • Tweezers, replaceable tip, 5 star quality
    Peek tips
    125 mm (5")
    0.7 mm (0.03")
    Straight Fine Replaceable points
    0.03" (0.7 mm)
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $241.45
    $241.45
    Each
       $241.45
    Supplier Number: 162B-RT
  • Tweezers, replaceable tip, 5 star quality
    Peek tips
    125 mm (5")
    1 mm (0.04")
    Straight Strong Replaceable Points
    0.04" (1 mm)
    Straight
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $227.40
    $227.40
    Each
       $227.40
    Supplier Number: 162C-RT
  • Tweezers, replaceable tip, 5 star quality
    Peek tips
    125 mm (5")
    0.3 mm (0.012")
    0.02" (0.5 mm)
    50°
    Discontinued
    Supplier Number: -
Replacement tips
Informations sur le produit
  • Catalog #
    Tip Width
    Tip Length
    Curvature
    Matière
    Désignation
    Availability
    Price
  • 0.02" (0.5 mm)
    0.3 mm (0.012")
    50°
    PEEK
    Replacement tips for 762a-rt, 5 star quality
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $95.12
    $95.12
    Each
       $95.12
Informations sur le produit
  • Catalog #
    Tip Width
    Tip Length
    Curvature
    Matière
    Désignation
    Availability
    Price
  • 0.08" (2 mm)
    0.4 mm (0.016")
    Straight
    PEEK
    Replacement tips for 162d-rt, 5 star quality
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.61
    $87.61
    Each
       $87.61
Informations sur le produit
  • Catalog #
    Tip Width
    Tip Length
    Curvature
    Matière
    Désignation
    Availability
    Price
  • 0.02" (0.5 mm)
    0.5 mm (0.02")
    Straight
    PEEK
    Replacement tips for 162a-rt, 5 star quality
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $122.33
    $122.33
    Each
       $122.33
Informations sur le produit
  • Catalog #
    Tip Width
    Tip Length
    Curvature
    Matière
    Désignation
    Availability
    Price
  • 0.03" (0.7 mm)
    0.7 mm (0.03")
    Straight
    PEEK
    Replacement tips for 162b-rt, 5 star quality
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $124.93
    $124.93
    Each
       $124.93