Notrax® T17 Superfoam™ Floor Mattings, Justrite®
: Justrite
Superfoam™ is an extremely lightweight and durable anti-fatigue foam mat designed specifically for ease of handling and ergonomic comfort.
- Closed cell nitrile foam, does not absorb liquids
- Durable and abrasion resistant
- Solid for dry areas
- Perforated drainage holes to remove liquid and debris for worker platform
- Non-allergenic, silicone-free compound
- 45° beveled edges on all four sides
Made from a closed cell PVC Nitrile foam blend, Superfoam™ does not absorb liquids and is highly resistant to greases, oils, animal and vegetable fats. This allows it to be used in areas where occasional overspray and dripping occur, such as dishwashing stations and bar areas. Because Superfoam™ is lightweight, it is the perfect mat for use in areas where mats are regularly moved for cleaning like cooking lines, service counters, and food prep stations.
Material: Sponge/Rubber
Frequently Bought Together