Notrax® 447 Comfort-Eze™ Floor Matting, Justrite®
76310-916
- Color:Black
- Length:1829 cm (60 ft.)
- Thickness:9 mm (⅜")
- Cat. no.:76310-916
- Weight:20 lbs.
- Width:91 cm (3 ft.)
Constructed with a unique solid rubber design, Comfort-Eze™ matting stimulates worker comfort and promotes product longevity.
- Long-lasting rubber compound
- Rubber molded, anti-fatigue mat
- Durable rubber top surface easy to wipe clean using most cleaners and disinfectants
- Beveled edges on all 4 sides
- 1800 sq. ft. coverage area
Offset bubbles on both the top and bottom of the mat provide flex and create airflow resulting in maximum comfort for workers required to be on their feet for extended periods. Competitive products feature only a flat or hollowed bottom which reduces the comfort of the mat.
Although intended for use in dry environments, the top surface design is easy to wipe clean and the durable rubber compound is highly resistant to many cleaners and disinfectants. Occasional spills can be cleaned up quickly and easily. Molded beveled edges on all four sides eliminate trip hazards by allowing for easy access on or off the mat.