Notrax® 850 Niru® Cushion-Ease® GSII™ Floor Mattings, Justrite®
Fournisseur: NOTRAX USA, INC.
Certificats
À propos de cet article
Niru® Cushion-Ease® GSII™ is a high-performance modular anti-fatigue matting system with a drainage top anti-slip surface texture and a multi-nib backing for comfort and ergonomic support of the mat.
- Resilient rubber compound and unique multi-nib support design offers exceptional fatigue relief
- Large hole drainage design keeps moisture and debris away
- Easy to snap tog system for on-site custom work station platforms
- Nitrile provides highest level of resistance to chemicals, oils, and greases
- GSII™ offers the ultimate in durability, performance, and traction in harsh environments
- Compatible with any Cushion-Ease® family mats
Made of 100% nitrile rubber compound that provides the highest resistance to chemicals, cutting fluids, animal fats, oils, and greases. GSII™ has a silicon carbide grit surface for extreme traction in the harshest work environment. Large holes provide excellent drainage and aeration leaving a dry, clean work area. All Cushion-Ease® mats have a male/female interlocking system and are compatible with Notrax® M.D. Ramp System® for easy on-site custom configurations and trip-resistant platforms.
Material: Rubber/Grit
Frequently Bought Together