Notrax® 482 Bubble Trax™ Floor Mattings, Justrite®
Fournisseur: NOTRAX USA, INC.
Certificats
À propos de cet article
Bubble Trax® is a unique high performance anti-fatigue floor mat with a bubble top surface design providing extreme ergonomic comfort and durability.
- Heavy-duty vinyl bubble top surface combined with a dense closed cell PVC foam base
- UniFusion™ bond guaranteed for the life of the mat
- Redstop™ non-slip backing to reduce mat slippage
- Beveled edges on all 4 sides
- Certified by the NFSI (National Floor Safety Institute)
Bubble Trax® combines a heavy-duty vinyl top surface with a dense sponge backing utilizing UniFusion™ technology. This virtually eliminates the possibility of de-lamination. All Notrax® laminate floor mats come standard with RedStop™, a uniquely engineered non-slip backing technology that eliminates slipping and sliding on surfaces such as tile, wood, marble, and treated concrete.
Material: Laminate/Vinyl
Frequently Bought Together