Notrax® 425 Superfoam™ Revive RS Solid Floor Mattings, Justrite®
Supplier: NOTRAX USA, INC.
Certificates
About this item
Durable rubberized sponge mat designed for extreme ergonomic comfort.
- Closed cell foam blend does not absorb liquids
- Durable, lightweight rubber is easy to pick up and move
- Ergonomically designed to provide high level of rebound without bottoming out
- Non-allergenic, silicone-free compound
- Beveled edges on all four sides
Made from a completely closed cell foam blend, Superfoam™ Revive RS does not absorb liquids and is highly resistant to grease, oils, and animal and vegetable fats. At 0.75" thick this anti-fatigue mat is ideal for workers standing for long periods of time.
Material: Sponge/Rubber
Frequently Bought Together