Biox Pipette Fillers
Fournisseur: TWD Scientific
Certificats
À propos de cet article
These pipette fillers are easy to use and feature fast aspiration and dispensing speeds, meanwhile maintaining accuracy every time.
- Ergonomic design
- 8+ hour battery life
- Compatible with 0.1 - 100 ml pipettes
- Battery life indicators
- Fast aspiration and dispense speeds
- Light weight
Pipette fillers are compatable with glass or plastic pipettes and feature an AC adaptor.
Spécifications
- Catalog No:
- 76319-864
- 76319-866
- Model:
- Levo Plus
- Levo ME
- Dispensing speed:
- 25 ml in <5 s
- 25 ml in <7 s
- Battery type:
- Lithium-ion
- Lithium-ion