Biox Micro PettePlus Pipettes | Avantor
Biox Micro PettePlus Pipettes
Biox Micro PettePlus Pipettes
Supplier:  TWD Scientific
Certificates
Biox Micro PettePlus Pipettes
Biox Micro PettePlus Pipettes
4 Options Available Below
Product Details & Documents
MicroPette Plus mechanical pipettes are the ideal adjustable volume pipettes for all of your basic pipetting needs.
  • Ergonomic design for comfortable extended use
  • Easy-to-read display
  • Convenient one-handed operation
  • Easy to calibrate and maintain with included tool
  • Individual piston and tip cone assemblies allow for easy repair and maintenance
  • Fully autoclavable
  • Compatible with most standard pipette tips
  • 4 sizes available
Micropette plus features single-channel adjustable volume. Adjustable in various increments depending on size.
Certifications: The MicroPette Plus is calibrated in accordance with ISO 8655, and each pipette includes its own individual test certificate
Informations sur les accessoires: Pipette Stand is available to accommodate up to 6 pipettes (01-2306)
Product Family Options
Informations sur le produit
  • Désignation
    Volume
    Increments
    Availability
    Price
  • MicroPette Plus Pipette
    0.5 - 10 µl
    0.1 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $240.60
    $240.60
    Each
       $240.60
    Supplier Number: BX01-2101
  • MicroPette Plus Pipette
    10 - 100 µl
    1 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $247.35
    $247.35
    Each
       $247.35
    Supplier Number: BX01-2104
  • MicroPette Plus Pipette
    20 - 200 µl
    1 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $240.60
    $240.60
    Each
       $240.60
    Supplier Number: BX01-2102
  • MicroPette Plus Pipette
    100 - 1000 µl
    5 µl
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $228.57
    $228.57
    Prix promotionnel pour les nouveaux clients
    Each
       $228.57
    Supplier Number: BX01-2110