Biologix dPette Digital Pipettes
These high-quality digital pipettes feature built in error control and follow a comprehensive range of liquid handling protocols.
- Three aspiration/ dispensing speeds
- LCD screen
- Motor drive with built in error control
- Lithium-ion rechargable battery
- Autoclavable lower barrel
dPette digital pipette features three dispening speeds and single-channel adjustable volume. Various sizes available.
: Pipette Stand is available to accommodate up to 6 pipettes (01-2306)
: Charging cord, charging base and power box included.Quality Control Certification Included.
Frequently Bought Together