VWR® Upright Freezer Racks for 25-Place Slide Boxes
Fournisseur: VWR International
Certificats
À propos de cet article
Designed for use with 25-place slide boxes.
- Made of corrosion resistant stainless steel
Security lock devices are available for each rack.
Frequently Bought Together