Document/Folder Holder, S-Curve
DH-1012 Wall mountable document holder, PETG, Clear, 12×10×2"
Catalog # 10693-202
Supplier: S-Curve Technologies
Supplier #: DH-1012
Jump to:
Product Details & Documents
Designed to hold cleanroom documents, papers, folders, and more.
Made from 1/8" thick clear PETG material.
Specifications
- Color:Clear
- Désignation:DH-1012 Wall mountable document holder
- Dimensions:12×10×2"
- Matière:PETG