Wiper Dispenser, S-Curve
Catalog # 10703-488
Supplier: S-Curve Technologies
Specifications
- Description:WD-050 Wiper Dispenser, with Lid
- Material:PETG
- Cat. no.:10703-488
- For Use With:5×5" Wipers
- Supplier No.:WDL-050PETG
Specifications
About this item
Clear PETG dispenser with lid. Designed for 5″ x 5″ wipes.
- Front opening allows better access to wipes
- Clear chemical resistant PETG material
- Wall mountable or free standing
- Flame polished edges for a smooth finish