Pipette Storage Container, Vertical, S-Curve Technologies
Catalog # 10703-474
Supplier: S-Curve Technologies
Specifications
- Description:PS-410 Pipette Storage Container with four compartments
- Dimensions:5.5W×23H×10"D
- Cat. no.:10703-474
- Supplier No.:PS-410
Specifications
About this item
Organize and protects pipettes.
- Clear 1/4" acrylic is durable and clean
- Flame polished edges for smoothness