Apparel Dispenser, Solid Front with Lid, S-Curve
Catalogue # 10693-116
Fournisseur: S-Curve Technologies
Spécifications
- Dispenser type:Floor-standing/wall-mountable
- Material:PETG
- No. of compartments:1
- PPE type:Garments
- Description:AD-001 garment dispenser, PETG
- Cat. no.:10693-116
- W×D×H:16×11×24"
- Supplier No.:AD-001LPETG
Spécifications
À propos de cet article
This solid front design keeps packaged and folded apparel organized and clean.
- Closed front model
- Mounts to wall or free standing
- Flame treated edges for a smooth finish
- Lid for better contamination control
Designed to hold coveralls, lab coats, frocks, smocks, aprons, and more.