Vikan® Large Hand Brushes, Stiff, Remco
Supplier: Remco Products
Certificates
About this item
This fully color-coded hand scrub brush has stiff angled bristles to scrub tables, conveyor, belts, cutting boards, buckets, and many other types of equipment.
- Stiff bristles made from polyester
- Polypropylene block
- Dishwasher safe
- Autoclavable up to 250°F
At one end of the brush, you have harder bristles, which help remove stubborn dirt.
Certifications: Meet FDA standards.
Frequently Bought Together