Macro Open Bottom Fluorescence Flow Through Cells, FireflySci
Marketsource
Macro Open Bottom Fluorescence Flow Through Cells, FireflySci
Marketsource
Macro Open Bottom Fluorescence Flow Through Cells, FireflySci
Macro Open Bottom Fluorescence Flow Through Cells, FireflySci
High-quality macro open bottom fluorescence flow through are of a great value.
  • Comes in several different materials: (Optical Glass, UV quartz, IR quartz)
  • Available in 10 mm lightpath
  • Four windows optically polished
  • Both ends are open
Fire-fused to ensure that cuvettes are robust and durable. Highly accurate results, only need to test sample once. These cuvettes are made with the best quality quartz and glass material available. Excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects. Each cell goes through a rigorous inspection process before leaving the factory.
Product Family Options
  • Volume
    Internal Dimensions
    External Dimensions
    Pathlength
    Matière
  • 4.000 ml
    L×W×H 10×10×40 mm
    L×W×H 12.5×12.5×40.0 mm
    10 mm
    Optical Glass
    Each
    Each - $724.80
    $724.80
    Each
       $724.80
     : 3FTG10
  • 4.000 ml
    L×W×H 10×10×40 mm
    L×W×H 12.5×12.5×40.0 mm
    10 mm
    IR Quartz
    Each
    Each - $678.44
    $678.44
    Each
       $678.44
     : 3FTIR10
    Marketsource
    Marketsource
  • 4.000 ml
    L×W×H 10×10×40 mm
    L×W×H 12.5×12.5×40.0 mm
    10 mm
    UV Quartz
    Each
    Each - $631.68
    $631.68
    Each
       $631.68
     : 3FTUV10
    Marketsource
    Marketsource