Screw caps for Microcentrifuge Tubes, Globe Scientific
These screwcaps for microcentrifuge tubes are suitable for all Globe Scientific screw top microcentrifuge tubes.
- Made of polypropylene
- Nitrile o-ring
The caps are available in a variety of colors and are offered either with or without an o-ring.
Frequently Bought Together