Pipette Dropping Clear Glass Bottles
Fournisseur: Wards
Certificats
À propos de cet article
Ideal for labs and classrooms in chemistry applications, these bottles are also great for storing, mixing, and dispensing essential oils.
- Non-screw top allows for quick and easy withdrawal
These borosilicate glass bottles come equipped with an eye dropper pipet for clean dispensing of the stored contents. The clear glass allows for maximum visibility and display of contents.
Frequently Bought Together