Buchner Filtering Flasks
Supplier: Wards
Jump to:
À propos de cet article
Buchner filtering flasks are made of high quality, heavy duty borosilicate glass.
Ordering information: Stopper not included.
8 Options Available Below
Product Details & Documents
Buchner filtering flasks are made of high quality, heavy duty borosilicate glass.
Ordering information: Stopper not included.
Product Family Options
Informations sur le produit
- DésignationCapacitéDiameter ×HAvailabilityPrice
- Supplier Number: CH0422ASpécifications:
More
Specifications
Cat. No.470313-976 - Supplier Number: CH0422BSpécifications:
More
Specifications
Cat. No.470313-978 - Supplier Number: CH0422CSpécifications:
More
Specifications
Cat. No.470313-980 - Supplier Number: CH0422C01Spécifications:
More
Specifications
Cat. No.470313-982 - Supplier Number: CH0422DSpécifications:
More
Specifications
Cat. No.470313-984 - Supplier Number: CH0422D01Spécifications:
More
Specifications
Cat. No.470313-986 - Supplier Number: CH0422ESpécifications:
More
Specifications
Cat. No.470313-988 - Supplier Number: CH0422FSpécifications:
More
Specifications
Cat. No.470313-990