The brand-new phenolic science lab tables will have perfect chemistry with any science classroom. These paragons of endurance are built with strong ash frame and side panel aprons beneath a solid, black, phenolic resin tabletop with a matte finish.
- Solid ash legs and apron
- Rubber liquid protecting boots
- Adjustable glides for accurate leveling
- Black, 0.75" phenolic top are highly impervious to most chemicals, acids, corrosions, stains and moisture
- Comparable to epoxy resin tops
- 10-year warranty
The work surface is crack-resistant and can be modified using carbide-tipped tools. It is impervious to extreme cold, heat up to 275 °F as well as damage from virtually any chemical, acid or possibly corrosive substance that may be found in a laboratory setting.
Robust legs are spot-welded to the frame with metal-on-metal double-bolts on 13-gauge steel corner braces for maximal stability. The PSLT's feet are outfitted with rubber liquid-protective boots and adjustable glides for accurate floor leveling.
Meets or exceeds SEFA environmental industrial standards.