Flow Through Cells with Side Tubes, FireflySci
Marketsource
Supplier:   FIREFLYSCI, INC.
Flow Through Cells with Side Tubes, FireflySci
Supplier:  FIREFLYSCI, INC.
Marketsource
Certificates
Flow Through Cells with Side Tubes, FireflySci
Flow Through Cells with Side Tubes, FireflySci
14 Options Available Below
Product Details & Documents
High-quality flow through cells in a variety of material types and are a great value.
  • Comes in several different materials including UV quartz and IR quartz
  • Two windows polished
  • Inlet/Outlet tubes: 5O.D.×3I.D.×15L mm
  • Height is without tubes
  • Lightpaths from 0.1 to 2 mm available
  • Tubes are made of quartz
  • Oval-shaped flow channel
Firefly's Type 48 flow through cell with sides tubes enables a constant flow through the observation chamber. Highly accurate results, only need to test sample once. Firefly cuvettes are made with the best quality quartz and glass material available. Excellent transmission values. Made by hand, ensuring high quality optical polishing with no defects.
Each cell goes through a vigorous inspection process before leaving the factory. Fire-fused to ensure that cuvettes are robust and durable.
 : Meets ASTM standards. EN 342
 : Accessories not included.
Product Family Options
Product Information
  • Volume
    Internal Dimensions
    External Dimensions
    Pathlength
    Matière
    Availability
    Price
  • 15 μl
    L×W×H 0.05×9.0×38.0 mm
    L×W×H 2.6×12.5×45.0 mm
    0.05 mm
    IR Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,568.81
    $1,568.81
    Each
       $1,568.81
    Supplier Number: 48IR0.05
    Specifications:
    Marketsource
    Marketsource
  • 30 µl
    L×W×H 0.10×9.0×38.0 mm
    L×W×H 2.6×12.5×45.0 mm
    0.1 mm
    IR Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,018.67
    $1,018.67
    Each
       $1,018.67
    Supplier Number: 48IR0.1
    Specifications:
    Marketsource
    Marketsource
  • 150 μl
    L×W×H 0.50×9.0×38.0 mm
    L×W×H 2.6×12.5×45.0 mm
    0.5 mm
    IR Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,018.67
    $1,018.67
    Each
       $1,018.67
    Supplier Number: 48IR0.5
    Specifications:
    Marketsource
    Marketsource
  • 300 μl
    L×W×H 1.00×9.0×38.0 mm
    L×W×H 3.5×12.5×45.0 mm
    1 mm
    IR Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,018.67
    $1,018.67
    Each
       $1,018.67
    Supplier Number: 48IR1
    Specifications:
    Marketsource
    Marketsource
  • 600 μl
    L×W×H 2.00×9.0×38.0 mm
    L×W×H 4.5×12.5×45.0 mm
    2 mm
    IR Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,018.67
    $1,018.67
    Each
       $1,018.67
    Supplier Number: 48IR2
    Specifications:
    Marketsource
    Marketsource
  • 1500 μl
    L×W×H 5.00×9.0×38.0 mm
    L×W×H 7.5×12.5×45.0 mm
    5 mm
    IR Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,018.67
    $1,018.67
    Each
       $1,018.67
    Supplier Number: 48IR5
    Specifications:
    Marketsource
    Marketsource
  • 3000 µl
    L×W×H 10.00×9.0×38.0 mm
    L×W×H 12.5×12.5×45.0 mm
    10 mm
    IR Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,018.67
    $1,018.67
    Each
       $1,018.67
    Supplier Number: 48IR10
    Specifications:
    Marketsource
    Marketsource
  • 15 μl
    L×W×H 0.05×9.0×38.0 mm
    L×W×H 3.0×12.5×45.0 mm
    0.05 mm
    UV Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,483.94
    $1,483.94
    Each
       $1,483.94
    Supplier Number: 48UV0.05
    Specifications:
    Marketsource
    Marketsource
  • 30 µl
    L×W×H 0.10×9.0×38.0 mm
    L×W×H 2.6×12.5×45.0 mm
    0.1 mm
    UV Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,002.92
    $1,002.92
    Each
       $1,002.92
    Supplier Number: 48UV0.1
    Specifications:
    Marketsource
    Marketsource
  • 150 μl
    L×W×H 0.50×9.0×38.0 mm
    L×W×H 2.6×12.5×45.0 mm
    0.5 mm
    UV Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,301.60
    $1,301.60
    Each
       $1,301.60
    Supplier Number: 48UV0.5
  • 300 μl
    L×W×H 1.00×9.0×38.0 mm
    L×W×H 3.5×12.5×45.0 mm
    1 mm
    UV Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,301.60
    $1,301.60
    Each
       $1,301.60
    Supplier Number: 48UV1
  • 600 μl
    L×W×H 2.00×9.0×38.0 mm
    L×W×H 4.5×12.5×45.0 mm
    2 mm
    UV Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,002.92
    $1,002.92
    Each
       $1,002.92
    Supplier Number: 48UV2
    Specifications:
    Marketsource
    Marketsource
  • 1500 μl
    L×W×H 5.00×9.0×38.0 mm
    L×W×H 7.5×12.5×45.0 mm
    5 mm
    UV Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,002.92
    $1,002.92
    Each
       $1,002.92
    Supplier Number: 48UV5
    Specifications:
    Marketsource
    Marketsource
  • 3000 µl
    L×W×H 10.00×9.0×38.0 mm
    L×W×H 12.5×12.5×45.0 mm
    10 mm
    UV Quartz
    Log in for availability
    Log in for availability
    Each
    Each - $1,002.92
    $1,002.92
    Each
       $1,002.92
    Supplier Number: 48UV10
    Specifications:
    Marketsource
    Marketsource