Type 18 Standard Micro Cuvettes with PTFE Cover, FireFlySci
These cuvettes are standard micro spectrophotometer cuvette with PTFE Cover.
- Highly accurate results, only need to test sample once
- Made by hand, ensuring high quality optical polishing with no defects
- Fire-fused to ensure that cuvettes are robust and durable
- Comes in several different materials: (Optical Glass, UV Quartz and IR Quartz)
- Lightpaths from 5 to 100 mm available
- Two windows optically polished
The narrower walls enables smaller sample sizes. Firefly cuvettes are made with high quality quartz and glass material available. High transmission values.
Each cell goes through a rigorous inspection process before leaving the factory.
: Includes PTFE cover.
Frequently Bought Together