Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Marketsource
Supplier:   FIREFLYSCI, INC.
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Supplier:  FIREFLYSCI, INC.
Marketsource
Certificates
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
Shimadzu Type 16 Multi-Micro Cell Array, FireflySci
2 Options Available Below
Product Details & Documents
Multi-Micro Cell Array has 16-chambered multi-micro array of cells for doing multiple measurements.
  • Highly accurate results, only need to test sample once
  • Made by hand, ensuring high quality optical polishing with no defects
  • Fire-fused to ensure that cuvettes are robust and durable
  • Comes in UV Quartz
  • Available in 10 mm lightpath
  • Two windows optically polished
  • Walls are made from black quartz to block out stray light
Typically used in Shimadzu machines.
Each cell goes through a rigorous inspection process before leaving the factory.
Product Family Options
Product Information
  • Désignation
    Volume
    Lightpath
    Availability
    Price
  • UV Quartz
    50 µl (16x)
    5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,021.36
    $3,021.36
    Each
       $3,021.36
    Minimum order quantity is 1
    Supplier Number: 16UV5
    Specifications:
    Marketsource
    Marketsource
  • UV Quartz
    100 µl (16×)
    10 mm
    Log in for availability
    Log in for availability
    Each
    Each - $3,021.36
    $3,021.36
    Each
       $3,021.36
    Minimum order quantity is 1
    Supplier Number: 16UV10
    Specifications:
    Marketsource
    Marketsource