Fixative for hematology, Solution D, (Wescor)
Hematology Stain, Solution D, Fixative (Wescor), 12 × 16 oz.
Catalog # CA10018-834
Supplier: Azer Scientific
Supplier #: ES946
Fixative for hematology, Solution D, (Wescor)
Catalog # CA10018-834
Supplier: Azer Scientific
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Compatible with the ELITechGroup Aerospray® stainer.
- Ready to use
- Lot to lot consistency
A Methanol fixative.
Specifications
Specifications
- Désignation:Hematology Stain, Solution D, Fixative (Wescor)
- Taille:12 × 16 oz.