Macro Cuvettes, Type 11, FireFlySci
Macro Cuvettes, Type 11, FireFlySci
Marketsource
Supplier:   FIREFLYSCI, INC.
Macro Cuvettes, Type 11, FireFlySci
Supplier:  FIREFLYSCI, INC.
Marketsource
Certificates
Macro Cuvettes, Type 11, FireFlySci
Macro Cuvettes, Type 11, FireFlySci
21 Options Available Below
Product Details & Documents
FireflySci's Type 11 cuvette comes standard with a glass cap, which provides an airtight seal, ensured by the ground finish on the inside of the cap and the top of the cuvette.
  • Hand made ensuring the highest quality optical polish with no defects
  • Firefly cuvettes are guaranteed to give high optical transmission
  • Fire fused to ensure the best bonding
  • Several different materials available: (Optical Glass, UV Quartz, IR Quartz)
  • Lightpaths from 1 mm up to 50 mm available
  • Two windows optically polished
The cap is polished, which enables optical transmission.
Every cell is thoroughly inspected before leaving the factory.
Delivery information: Includes airtight glass cap.
Product Family Options
Optical Glass
Informations sur le produit
  • Volume
    Lightpath
    Cap Type
    Availability
    Price
  • 0.40 ml
    1 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,027.33
    $1,027.33
    Each
       $1,027.33
    Minimum order quantity is 1
    Supplier Number: 11G1
    Spécifications:
    Marketsource
    Marketsource
  • 0.70 ml
    2 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,042.94
    $1,042.94
    Each
       $1,042.94
    Minimum order quantity is 1
    Supplier Number: 11G2
    Spécifications:
    Marketsource
    Marketsource
  • 1.70 ml
    5 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $816.83
    $816.83
    Each
       $816.83
    Minimum order quantity is 1
    Supplier Number: 11G5
    Spécifications:
    Marketsource
    Marketsource
  • 3.50 ml
    10 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $757.94
    $757.94
    Each
       $757.94
    Minimum order quantity is 1
    Supplier Number: 11G10
    Spécifications:
    Marketsource
    Marketsource
  • 7.00 ml
    20 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $783.17
    $783.17
    Each
       $783.17
    Minimum order quantity is 1
    Supplier Number: 11G20
    Spécifications:
    Marketsource
    Marketsource
  • 14.0 ml
    40 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $934.06
    $934.06
    Each
       $934.06
    Minimum order quantity is 1
    Supplier Number: 11G40
    Spécifications:
    Marketsource
    Marketsource
  • 17.5 ml
    50 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,068.13
    $1,068.13
    Each
       $1,068.13
    Minimum order quantity is 1
    Supplier Number: 11G50
    Spécifications:
    Marketsource
    Marketsource
Infrared Quartz
Informations sur le produit
  • Volume
    Lightpath
    Cap Type
    Availability
    Price
  • 0.40 ml
    1 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,637.63
    $1,637.63
    Each
       $1,637.63
    Supplier Number: 11IR1
  • 0.70 ml
    2 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,260.68
    $1,260.68
    Each
       $1,260.68
    Minimum order quantity is 1
    Supplier Number: 11IR2
    Spécifications:
    Marketsource
    Marketsource
  • 1.70 ml
    5 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,080.63
    $1,080.63
    Each
       $1,080.63
    Minimum order quantity is 1
    Supplier Number: 11IR5
    Spécifications:
    Marketsource
    Marketsource
  • 3.50 ml
    10 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $980.07
    $980.07
    Each
       $980.07
    Minimum order quantity is 1
    Supplier Number: 11IR10
    Spécifications:
    Marketsource
    Marketsource
  • 7.00 ml
    20 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,084.84
    $1,084.84
    Each
       $1,084.84
    Minimum order quantity is 1
    Supplier Number: 11IR20
    Spécifications:
    Marketsource
    Marketsource
  • 14.0 ml
    40 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,197.92
    $1,197.92
    Each
       $1,197.92
    Minimum order quantity is 1
    Supplier Number: 11IR40
    Spécifications:
    Marketsource
    Marketsource
  • 17.5 ml
    50 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,323.57
    $1,323.57
    Each
       $1,323.57
    Minimum order quantity is 1
    Supplier Number: 11IR50
    Spécifications:
    Marketsource
    Marketsource
Ultraviolet Quartz
Informations sur le produit
  • Volume
    Lightpath
    Cap Type
    Availability
    Price
  • 0.40 ml
    1 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,553.95
    $1,553.95
    Each
       $1,553.95
    Supplier Number: 11UV1
  • 0.70 ml
    2 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,197.92
    $1,197.92
    Each
       $1,197.92
    Minimum order quantity is 1
    Supplier Number: 11UV2
    Spécifications:
    Marketsource
    Marketsource
  • 1.70 ml
    5 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,315.15
    $1,315.15
    Each
       $1,315.15
    Supplier Number: 11UV5
  • 3.50 ml
    10 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $917.30
    $917.30
    Each
       $917.30
    Minimum order quantity is 1
    Supplier Number: 11UV10
    Spécifications:
    Marketsource
    Marketsource
  • 7.00 ml
    20 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,009.51
    $1,009.51
    Each
       $1,009.51
    Minimum order quantity is 1
    Supplier Number: 11UV20
    Spécifications:
    Marketsource
    Marketsource
  • 14.0 ml
    40 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,109.95
    $1,109.95
    Each
       $1,109.95
    Minimum order quantity is 1
    Supplier Number: 11UV40
    Spécifications:
    Marketsource
    Marketsource
  • 17.5 ml
    50 mm
    Glass cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,306.71
    $1,306.71
    Each
       $1,306.71
    Minimum order quantity is 1
    Supplier Number: 11UV50
    Spécifications:
    Marketsource
    Marketsource