Deflagration Spoon | Avantor
Deflagration Spoon
Deflagration Spoon
Catalog # 470313-336
Supplier:   GSC International
Fournisseur #:  4-32901-12
Deflagration Spoon
Catalog # 470313-336
Supplier:  GSC International
Deflagration Spoon
Deflagration Spoon
Deflagration Spoon
Price
$17.20
Log in pour la disponibilité
Each Pack of 12
La quantité minimale de commande est de 12
Product Details & Documents
Deflagration Spoons are commonly used in experiments that require students to burn phosphorous, sulfur, or other substances at a safe distance.
The handle is angled to keep hands away from the heat source. It is 18" long with a 3/4" diameter bowl and 1/4" depth.
Spécifications
  • Désignation:
    Deflagration Spoon