ContactUsLinkComponent
Spatulal and Spoon Tools
  470313-332
undefined
Spatulal and Spoon Tools
  470313-332
 :  4-18351-PK
  • Description:
    Spatula and spoon tool, 6"
  • Cat. no.:
    470313-332
This nickel-plated spoon and spatula tool is 14.9 cm long.
The bowl of its spoon measures 30 mm by 22 mm and has a 2 ml capacity, while the spatula measures 35 mm by 18 mm.