Acetate Buffer Solution, Fluka™, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
New Product
Certificates
About this item
This pH buffer solution is a combination of sodium acetate/acetic acid in 1:1 ratio for complexometry.
Frequently Bought Together