Ultraviolet Fluorometer Cuvettes, FireFlySci
Marketsource
Supplier: FIREFLYSCI, INC.
Jump to:
À propos de cet article
Ultraviolet Fluorometer Cuvettes have four windows with base polished and quartz cap.
4 Options Available Below
Product Details & Documents
Ultraviolet Fluorometer Cuvettes have four windows with base polished and quartz cap.
Product Family Options
Informations sur le produit
- LightpathCap TypeAvailabilityPrice
- Supplier Number: 11FLUV5Spécifications:MarketsourceMarketsource
- Supplier Number: 11FLUV10
- Supplier Number: 11FLUV20Spécifications:MarketsourceMarketsource
- Supplier Number: 11FLUV40Spécifications:MarketsourceMarketsource