Quanta-Ray Dye Laser Flow Through Cells, FireFlySci
Catalog # 76015-076
Supplier: FIREFLYSCI, INC.
Specifications
- Description:Dye Laser Flow Through Cell
- Material:UV Quartz
- Lightpath:6×12 mm
- Cat. no.:76015-076
Specifications
About this item
Dye laser cells are manufactured with extreme surface flatness extending to the edges of the cell.
- Highly accurate results, only need to test sample once
- Made by hand, ensuring high quality optical polishing with no defects
- Each cell goes through a rigorous inspection process before leaving our factory
- Fire-fused to ensure that cuvettes are robust and durable
- Available in UV Quartz
- Available in dual 6×12 mm lightpath
- Four windows optically polished
- Open top and open bottom flow through cell
Flatness is checked via interferometer. Premium quality Firefly cuvettes are made with the best quality quartz and glass material available transmission values.