Tissue Marking Dyes, Azer Scientific
Mark critical margins and areas of interest on tissue sections with these assorted colors of tissue marking dye.
- Available in black and six vibrant colors
- Colors: black, blue, green, orange, red, violet, yellow
- Saturated color quickly tranfers to tissue
- Ready-to-use formula
Mark margins and areas of interest on tissue samples with this tissue marking dye.
: Dyes are available individually in 2 oz. and 8 oz. bottles.
: Kits include 2 oz. bottles of each color dye, a base and applicator sticks.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together