Light Shielding Extension Lines, SAI Infusion Technologies
Fournisseur: SAI Infusion Technologies
Certificats
À propos de cet article
Protect light-sensitive compounds during dosing with these purpose-built, light-shielding polyurethane extension lines.
- One male and one female Luer
- Can be customized to fit user needs
- Supplied sterile
- For laboratory animal use only
These radiopaque tubings eliminates the headaches and mistakes associated with covering lines by hand. Custom sizes are available in both black and clear lines.
Packaging: 10 per pack.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together