Trays and Inserts, Gratnells
Supplier:   GRATNELLS LLC
Trays and Inserts, Gratnells
Supplier:  GRATNELLS LLC
Certificates
Trays and Inserts, Gratnells
Trays and Inserts, Gratnells
75 Options Available Below
Product Details & Documents
These all adds up to a comprehensive storage system.
  • Available in a range of shapes and sizes
  • Available in many colors
  • Rigid and foam inserts available
  • Lid available to cover contents
These award-winning trays come in a range of shapes and sizes to fit most standard school furniture. Made from tough, recyclable polypropylene, these trays will last for many years. Standard F trays are available in a wide variety of colors to suit most color schemes. These F trays are designed to fit 450 mm deep furniture. Available in shallow, deep, extra deep and jumbo size.
If you need to keep stored items covered, the easy Snap-On lids help to keep contents secure. Keep contents organized with the comprehensive range of tray inserts for storing smaller items such as pens or crayons with three, four, six, eight or twelve separate compartments in each insert. Foam inserts are also available, which are ideal for storing calculators and science equipment.
In order to safely store smaller items, a wide variety of molded polystyrene vacuum formed inserts which fit in the F series trays. A variety of different molded compartments and sizes are available. The one section insert is a black plastic insert that sits at the top of the tray allowing a second insert on the tray base. Lids still fit with the one section insert. This one section inserts are available in three different depths.
A range of foam inserts for the F series trays hold delicate equipment safely and store a range of differently shaped items with ease. Blank foams are also available in a variety of depths.
Product Family Options
Shallow (F1) Trays
Informations sur le produit
  • Désignation
    Color
    Volume
    Exterior Dimensions
    Interior Dimensions
    Availability
    Price
  • Sunshine Yellow
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22.90
    $22.90
    Log in pour la disponibilité
    Pallet of 256
    Pallet of 256 - $5,838.10
    $5,838.10
    Each
       $22.90
    Pallet of 256
       $5,838.10
    Supplier Number: F0102
  • Sunshine Yellow
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $239.40
    $239.40
    Pack of 1
       $239.40
    Supplier Number: F0102P8
  • Plum Purple
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22.90
    $22.90
    Log in pour la disponibilité
    Pallet of 256
    Pallet of 256 - $5,838.10
    $5,838.10
    Each
       $22.90
    Pallet of 256
       $5,838.10
    Supplier Number: F0105
  • Plum Purple
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 8
    Pack of 8 - $239.40
    $239.40
    Pack of 8
       $239.40
    Supplier Number: F0105P8
  • Royal Blue
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22.90
    $22.90
    Log in pour la disponibilité
    Pallet of 256
    Pallet of 256 - $5,838.10
    $5,838.10
    Each
       $22.90
    Pallet of 256
       $5,838.10
    Supplier Number: F0106
  • Royal Blue
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $248.35
    $248.35
    Pack of 1
       $248.35
    Supplier Number: F0106P8
  • Flame Red
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.65
    $11.65
    Liquidation
    Log in pour la disponibilité
    Pallet of 256
    Pallet of 256 - $2,976.50
    $2,976.50
    Liquidation
    Each
       $11.65
    Pallet of 256
       $2,976.50
    Supplier Number: F0109
  • Flame Red
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $239.40
    $239.40
    Pack of 1
       $239.40
    Supplier Number: F0109P8
  • Grass Green
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.65
    $11.65
    Liquidation
    Log in pour la disponibilité
    Pallet of 256
    Pallet of 256 - $2,976.50
    $2,976.50
    Liquidation
    Each
       $11.65
    Pallet of 256
       $2,976.50
    Supplier Number: F0110
  • Grass Green
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $239.40
    $239.40
    Pack of 1
       $239.40
    Supplier Number: F0110P8
  • Translucent
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22.90
    $22.90
    Log in pour la disponibilité
    Pallet of 256
    Pallet of 256 - $5,838.10
    $5,838.10
    Each
       $22.90
    Pallet of 256
       $5,838.10
    Supplier Number: F0120
  • Translucent
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $231.65
    $231.65
    Pack of 1
       $231.65
    Supplier Number: F0120P8
  • Lime Jolly
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $11.65
    $11.65
    Liquidation
    Log in pour la disponibilité
    Pallet of 256
    Pallet of 256 - $2,976.50
    $2,976.50
    Liquidation
    Each
       $11.65
    Pallet of 256
       $2,976.50
    Supplier Number: F0136
  • Lime Jolly
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $239.40
    $239.40
    Pack of 1
       $239.40
    Supplier Number: F0136P8
  • Cyan Blue
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $239.40
    $239.40
    Pack of 1
       $239.40
    Supplier Number: F0126P8
  • Tropical Orange
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $239.40
    $239.40
    Pack of 1
       $239.40
    Supplier Number: F0101P8
  • Light Gray
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $247.85
    $247.85
    Pack of 1
       $247.85
    Supplier Number: F0119P8
  • Silver
    7.5 L
    75H×312W×427L mm
    70H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $239.40
    $239.40
    Pack of 1
       $239.40
    Supplier Number: F0127P8
Shallow (F1) Tray Inserts
Informations sur le produit
  • Désignation
    Color
    Volume
    Exterior Dimensions
    Interior Dimensions
    Availability
    Price
  • Discontinued
    Supplier Number: -
  • 1 Large and 2 Small Sections
    Dove Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $149.45
    $149.45
    Pack of 6
       $149.45
    Supplier Number: IM02F0119P6
  • Three Section Insert
    Dove Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $149.45
    $149.45
    Pack of 1
       $149.45
    Supplier Number: IM03F0119P6
  • Four Section Insert
    Dove Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $149.45
    $149.45
    Pack of 6
       $149.45
    Supplier Number: IM04F0119P6
  • Six Section Insert
    Dove Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $149.45
    $149.45
    Pack of 1
       $149.45
    Supplier Number: IM06F0119P6
  • 6 Non Identical Sections
    Dove Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $149.45
    $149.45
    Pack of 1
       $149.45
    Supplier Number: IM07F0119P6
  • Eight Section Insert
    Dove Gray
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $149.45
    $149.45
    Pack of 1
       $149.45
    Supplier Number: IM08F0119P6
  • Crayon Insert 3 - 12 Compartments
    White
    Discontinued
    Supplier Number: -
Deep (F2) Trays
Informations sur le produit
  • Désignation
    Color
    Volume
    Exterior Dimensions
    Interior Dimensions
    Availability
    Price
  • Sunshine Yellow
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.95
    $15.95
    Liquidation
    Log in pour la disponibilité
    Pallet of 160
    Pallet of 160 - $2,550.25
    $2,550.25
    Liquidation
    Each
       $15.95
    Pallet of 160
       $2,550.25
    Supplier Number: F0202
  • Sunshine Yellow
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0202P6
  • Plum Purple
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.15
    $31.15
    Log in pour la disponibilité
    Pallet of 160
    Pallet of 160 - $4,977.00
    $4,977.00
    Each
       $31.15
    Pallet of 160
       $4,977.00
    Supplier Number: F0205
  • Plum Purple
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0205P6
  • Royal Blue
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.15
    $31.15
    Log in pour la disponibilité
    Pallet of 160
    Pallet of 160 - $4,977.00
    $4,977.00
    Each
       $31.15
    Pallet of 160
       $4,977.00
    Supplier Number: F0206
  • Royal Blue
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0206P6
  • Flame Red
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.95
    $15.95
    Liquidation
    Log in pour la disponibilité
    Pallet of 160
    Pallet of 160 - $2,550.25
    $2,550.25
    Liquidation
    Each
       $15.95
    Pallet of 160
       $2,550.25
    Supplier Number: F0209
  • Flame Red
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0209P6
  • Grass Green
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.95
    $15.95
    Liquidation
    Log in pour la disponibilité
    Pallet of 160
    Pallet of 160 - $2,550.25
    $2,550.25
    Liquidation
    Each
       $15.95
    Pallet of 160
       $2,550.25
    Supplier Number: F0210
  • Grass Green
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0210P6
  • Translucent
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.15
    $31.15
    Log in pour la disponibilité
    Pallet of 160
    Pallet of 160 - $4,977.00
    $4,977.00
    Each
       $31.15
    Pallet of 160
       $4,977.00
    Supplier Number: F0220
  • Translucent
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $255.90
    $255.90
    Pack of 6
       $255.90
    Supplier Number: F0220P6
  • Lime Jolly
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.95
    $15.95
    Liquidation
    Log in pour la disponibilité
    Pallet of 160
    Pallet of 160 - $2,550.25
    $2,550.25
    Liquidation
    Each
       $15.95
    Pallet of 160
       $2,550.25
    Supplier Number: F0236
  • Lime Jolly
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0236P6
  • Tropical Orange
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0201P6
  • Light Gray
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0219P6
  • Cyan Blue
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0226P6
  • Silver
    15 L
    150H×312W×427L mm
    140H×283W×385L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $255.90
    $255.90
    Pack of 1
       $255.90
    Supplier Number: F0227P6
Extra Deep (F25) Trays
Informations sur le produit
  • Désignation
    Color
    Volume
    Exterior Dimensions
    Interior Dimensions
    Availability
    Price
  • Translucent
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $23.65
    $23.65
    Liquidation
    Log in pour la disponibilité
    Pallet of 80
    Pallet of 80 - $1,891.10
    $1,891.10
    Liquidation
    Each
       $23.65
    Pallet of 80
       $1,891.10
    Supplier Number: F2520
  • Translucent
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2520P6
  • Tropical Orange
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2501P6
  • Sunshine Yellow
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $325.25
    $325.25
    Pack of 1
       $325.25
    Supplier Number: F2502P6
  • Plum Purple
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2505P6
  • Royal Blue
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2506P6
  • Flame Red
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2509P6
  • Grass Green
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2510P6
  • Light Gray
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2519P6
  • Cyan Blue
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2526P6
  • Silver
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $328.50
    $328.50
    Pack of 1
       $328.50
    Supplier Number: F2527P6
  • Lime Jolly
    22.5 L
    225H×312W×430L mm
    210H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $307.85
    $307.85
    Pack of 1
       $307.85
    Supplier Number: F2536P6
Jumbo (F3) Trays
Informations sur le produit
  • Désignation
    Color
    Volume
    Exterior Dimensions
    Interior Dimensions
    Availability
    Price
  • Sunshine Yellow
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $22.10
    $22.10
    Liquidation
    Log in pour la disponibilité
    Pallet of 72
    Pallet of 72 - $1,591.15
    $1,591.15
    Liquidation
    Each
       $22.10
    Pallet of 72
       $1,591.15
    Supplier Number: F0302
  • Sunshine Yellow
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0302P6
  • Plum Purple
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26.50
    $26.50
    Liquidation
    Log in pour la disponibilité
    Pallet of 72
    Pallet of 72 - $1,907.15
    $1,907.15
    Liquidation
    Each
       $26.50
    Pallet of 72
       $1,907.15
    Supplier Number: F0305
  • Plum Purple
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0305P6
  • Royal Blue
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26.50
    $26.50
    Liquidation
    Log in pour la disponibilité
    Pallet of 72
    Pallet of 72 - $1,907.15
    $1,907.15
    Liquidation
    Each
       $26.50
    Pallet of 72
       $1,907.15
    Supplier Number: F0306
  • Royal Blue
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0306P6
  • Flame Red
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26.50
    $26.50
    Liquidation
    Log in pour la disponibilité
    Pallet of 72
    Pallet of 72 - $1,907.15
    $1,907.15
    Liquidation
    Each
       $26.50
    Pallet of 72
       $1,907.15
    Supplier Number: F0309
  • Flame Red
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0309P6
  • Grass Green
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26.50
    $26.50
    Liquidation
    Log in pour la disponibilité
    Pallet of 72
    Pallet of 72 - $1,907.15
    $1,907.15
    Liquidation
    Each
       $26.50
    Pallet of 72
       $1,907.15
    Supplier Number: F0310
  • Grass Green
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0310P6
  • Translucent
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $51.80
    $51.80
    Log in pour la disponibilité
    Pallet of 72
    Pallet of 72 - $3,720.90
    $3,720.90
    Each
       $51.80
    Pallet of 72
       $3,720.90
    Supplier Number: F0320
  • Translucent
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0320P6
  • Lime Jolly
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $26.50
    $26.50
    Liquidation
    Log in pour la disponibilité
    Pallet of 72
    Pallet of 72 - $1,907.15
    $1,907.15
    Liquidation
    Each
       $26.50
    Pallet of 72
       $1,907.15
    Supplier Number: F0336
  • Lime Jolly
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0336P6
  • Tropical Orange
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0301P6
  • Light Gray
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0319P6
  • Cyan Blue
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0326P6
  • Silver
    30 L
    300H×312W×430L mm
    280H×276W×376L mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $386.70
    $386.70
    Pack of 1
       $386.70
    Supplier Number: F0327P6
Runners
Informations sur le produit
  • Désignation
    Color
    Volume
    Exterior Dimensions
    Interior Dimensions
    Availability
    Price
  • StopSafe Runner, 100 Pairs
    Translucent
    Discontinued
    Supplier Number: -
  • Tray Clips for StopSafe Runner, 100 Pairs
    Translucent
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $1,030.30
    $1,030.30
    Pack of 1
       $1,030.30
    Supplier Number: CCLIP19P100
  • Top Fixing Standard Runner, 12 mm, 6 Pairs
    White
    Discontinued
    Supplier Number: -
  • Top Fixing Standard Runner, 50 mm, 6 Pairs
    White
    Discontinued
    Supplier Number: -
Lids
Informations sur le produit
  • Désignation
    Color
    Volume
    Exterior Dimensions
    Interior Dimensions
    Availability
    Price
  • Clip On Lid for F Tray
    Translucent
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7.40
    $7.40
    Liquidation
    Log in pour la disponibilité
    Pallet of 460
    Pallet of 460 - $3,400.30
    $3,400.30
    Liquidation
    Each
       $7.40
    Pallet of 460
       $3,400.30
    Supplier Number: LF1
  • Clip On Lid for F Tray, Pack of 8
    Translucent
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1
    Pack of 1 - $126.25
    $126.25
    Pack of 1
       $126.25
    Supplier Number: LF1P8