ESD Safe Workstation Covers, Transforming Technologies
ESD Safe Workstation Covers, Transforming Technologies
ESD Safe Workstation Covers, Transforming Technologies
ESD Safe Workstation Covers, Transforming Technologies
Drape ESD workstation covers over sensitive products to protect from ESD, dust, and other contaminants.
  • Groundable for added safety; 4 mm grounding snap included
  • Polyester with conductive yarn
  • ESD-safe logo designates safe use in ESD protective environments
  • Resistance less than 10⁵
  • Bound edges
ESD workstation covers create a Faraday cage effect around the covered product to shield from electrostatic fields and electrostatic discharges (ESD). This washable cover has a surface resistivity of 10⁵ ohms/sq. Covers feature a silk-screened ESD symbol.
Product Family Options
  • Color
    W×L
  • Blue
    2×3'
    Each
    Each - $45.10
    $45.10
    Each
       $45.10
     : WC2436B
  • Blue
    4×3'
    Each
    Each - $90.20
    $90.20
    Each
       $90.20
     : WC4836B
  • Blue
    6×3'
    Each
    Each - $136.00
    $136.00
    Each
       $136.00
     : WC7236B