ProSolo Cable Options, ODO®/CT Cable Assemblies, YSI
ProSolo Cable Options, ODO®/CT Cable Assemblies, YSI
Supplier:   YSI
ProSolo Cable Options, ODO®/CT Cable Assemblies, YSI
Supplier:  YSI
Certificates
ProSolo Cable Options, ODO®/CT Cable Assemblies, YSI
ProSolo Cable Options, ODO®/CT Cable Assemblies, YSI
6 Options Available Below
Product Details & Documents
Connect any ProDIGITAL integrated probe/cable assembly the ProSolo to form a complete sampling system. Probe assemblies are user-replaceable with cable lengths ranging from 1 to 100 meters.
  • Dissolved oxygen technology has advanced from electrochemical sensors, such as polarographic and galvanic to luminescent-based optical sensors
  • Digital smart sensors
  • Single cable design for probe assembly lengths ranging from 1 to 100 meters
The ODO®/CT probe assemblies pair with the ProSolo Handheld to form a complete sampling system. The ODO®/CT has all the advantages of optical dissolved oxygen technology – higher accuracy, less maintenance, no stirring required for accurate measurements, holds calibration longer than membrane covered sensors and is not susceptible to interferences from other gases.
Optical Dissolved Oxygen (ODO®) technology is non-consumptive, meaning oxygen is not consumed during the measurement. Flow dependence is non-existent and no stirring is required, resulting in the most accurate DO measurements available from a probe. The ODO® sensor is not susceptible to interferences from other gases, drifts less and holds calibration longer than membrane covered sensors. Maintenance is a breeze with a simple, threaded optical cap that is good for at least two years.
Instrument setup is quick and easy with the integral sensors, which eliminates the need to install sensors before use. The digital technology allows the handheld to automatically recognize connected sensors, so there is no need to tell the instrument what is connected.
Ordering information: Available cable options; contact our Avantor representative for more information.
Delivery information: Two-year warranty on cable, probe, and ODO sensor cap.
Product Family Options
Product Information
  • Ø
    Cable Length
    Availability
    Price
  • 2.46 cm
    1 m
    Log in for availability
    Log in for availability
    Each
    Each - $2,941.00
    $2,941.00
    Each
       $2,941.00
    Supplier Number: 627150-1
  • 2.46 cm
    4 m
    Log in for availability
    Log in for availability
    Each
    Each - $2,940.98
    $2,940.98
    Each
       $2,940.98
    Minimum order quantity is 1
    Supplier Number: 627150-4
  • 2.46 cm
    10 m
    Log in for availability
    Log in for availability
    Each
    Each - $3,319.69
    $3,319.69
    Each
       $3,319.69
    Minimum order quantity is 1
    Supplier Number: 627150-10
  • 2.46 cm
    20 m
    Log in for availability
    Log in for availability
    Each
    Each - $3,385.00
    $3,385.00
    Each
       $3,385.00
    Minimum order quantity is 1
    Supplier Number: 627150-20
  • 2.46 cm
    30 m
    Discontinued
    Supplier Number: 627150-30
  • 2.46 cm
    50 m
    Log in for availability
    Log in for availability
    Each
    Each - $3,625.99
    $3,625.99
    Each
       $3,625.99
    Supplier Number: 627150-50
  • 2.46 cm
    100 m
    Log in for availability
    Log in for availability
    Each
    Each - $4,056.73
    $4,056.73
    Each
       $4,056.73
    Minimum order quantity is 1
    Supplier Number: 627150-100