À propos de cet article
Ward’s® PURE Preserved Specimens - the best dissection experience from start to finish.
- Safest specimens
- Low formalin odor
- Best classroom experience
Ward's® PURE Preserved Specimens offer the best classroom experience because PURE Specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
Dissection specimens that encourages discovery and real life scenarios. Learn areas of the brain, discuss ramifications of the tumor in your patient, and find your simulated tumor in this all new interactive product!
Sheep brains contain simulated tumors in three different areas of the brain: the hypothalamus, motor cortex, and visual cortex. Each set comes with either 2, 5, or 10 of each example.
Hypothalamus: The hypothalamus main role is to keep the body in homeostasis as much as possible. Examples include regulating body temperature, emotions, blood pressure and heart rate, appetite.
Occipital lobe: Controls visual functions.
Motor cortex: Involved in planning, control, and execution of voluntary movements.