Replacement Shield Plus Visor, Portwest
Replacement visor PW99, Clear, PC, 7.9×15.7"
Catalog # 76105-174
Supplier: Portwest
: PW99CLR
Jump to:
Product Details & Documents
Replacement visor for PW96. It also fits PW58 and PS58. Made from durable Polycarbonate with 1mm thickness.
- Clear visor aluminium bound
- For medium-energy impact protection
- Protects from impacts, metal splashes and hot solid projections
- Protection against Molten Metal
Replacement visor to be used on PW96 browguard support. Compatible with PW58 helmet visor carrier and PS58 helmet visor holder.
Spécifications
- Color:Clear
- Désignation:Replacement visor PW99
- Matière:PC
- Taille:7.9×15.7"